Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT-1 Rabbit Polyclonal Antibody, Epredia™

GLUT-1, Rabbit Polyclonal Ab is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Supplier:  Epredia RB9052P

 View more versions of this product

Catalog No. RB9052P


Only null left
Add to Cart
Request a Quote

Description

Description

Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon, lung, stomach, esophagus, and breast. GLUT-1 immunoreactivity in some cancers, including trans carcinoma of the urinary bladder, has been associated with aggressive behavior.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from C-terminal of human GLUT-1

Molecular Weight: 55 kDa, 45 kDa depending on the extent of glycosylation

Positive Control: Rat brain lysate; Breast CA, squamous cell CA, nonsmall-cell lung CA and esophageal CA

Cellular Localization: Membrane and focal cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
Specifications

Specifications

GLUT-1
Polyclonal
200µg/mL
Rabbit
1 mL
Steroids Hormones and Growth Factors
Human
IgG
Immunohistochemistry (Paraffin), Western Blot
Polyclonal
Unconjugated
A synthetic peptide derived from C-terminal of human GLUT-1
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.