Learn More
GLUT-1 Rabbit Polyclonal Antibody, Epredia™
GLUT-1, Rabbit Polyclonal Ab is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Supplier: Epredia RB9052R1
Description
Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon, lung, stomach, esophagus, and breast. GLUT-1 immunoreactivity in some cancers, including trans carcinoma of the urinary bladder, has been associated with aggressive behavior.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Rat. Others not tested.
Epitope: C-terminal
Immunogen: A synthetic peptide derived from C-terminal of human GLUT-1
Molecular Weight: 55 kDa, 45 kDa depending on the extent of glycosylation
Positive Control: Rat brain lysate; Breast CA, squamous cell CA, nonsmall-cell lung CA and esophageal CA
Cellular Localization: Membrane and focal cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Specifications
GLUT-1 | |
Polyclonal | |
Unconjugated | |
A synthetic peptide derived from C-terminal of human GLUT-1 | |
IVD | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin), Western Blot | |
Polyclonal | |
Rabbit | |
1 mL | |
Steroids Hormones and Growth Factors | |
Human | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.