Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Growth Hormone, Human/hGH, Ab-3 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1328R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1328R7 7 mL Prediluted Prediluted
MS1328P 1 mL Concentrated 200μg/mL
MS1328P0 100 μL Concentrated 200μg/mL
MS1328P1ABX 100 μL Concentrated 1mg/mL
MS1328PABX 200 μL Concentrated 1mg/mL
MS1328P1 500 μL Concentrated 200μg/mL
MS1328R1 1 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1328R7 Supplier Epredia Supplier No. MS1328R7

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments.

hGH is produced by the somatotroph cells in the pituitary. hGH acts through binding to membrane receptors that belong to the cytokine receptor superfamily. Ligand binding induces receptor dimerization and activation of the receptor-associated kinase: JAK2; this results in phosphorylation of the kinase itself, of the receptor, and of many cellular proteins including the Stat proteins as well as adaptors leading to the activation of the Ras/MAP kinase pathway and of the PI-3 kinase pathway.

Host Species: Mouse

Clone: GH01/#2 + GH02/#7

Isotype: IgG2a/κ + IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant human growth hormone

Molecular Weight: ∽22 kDa

Positive Control: Recombinant human growth hormone, Human anterior pituitary

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Growth Hormone, Human/hGH Ab-3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone GH01 / #2 + GH02 / #7
Concentration Prediluted
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified recombinant human growth hormone
Quantity 7 mL
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.