Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ GRP94 Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB10642P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB10642P 1 mL Concentrated
RB10642P0 100 μL Concentrated
RB10642P1 500 μL Concentrated
RB10642R1 1 mL Prediluted
RB10642R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RB10642P Supplier Epredia™ Supplier No. RB10642P

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ GRP94, Rabbit Polyclonal Antibody.

Heat shock protein (HSP) molecular chaperones are environmental stress inducible gene products. The human HSP 90 family consists of 17 genes classified into four classes: HSP90AA, HSP90AB, HSP90B and TRAP. HSP 90 family members guide the normal folding, intracellular disposition and proteolytic turnover of many key regulators of cell growth, differentiation and survival. GRP 94, glucose regulated protein 94 kDa, a HSP90 family member, (also known as tumor rejection antigen 1 (TRA1), ECGP and GP96), localizes to the ER, is highly expressed in BGC-823 human gastric carcinoma cells and is upregulated in human endothelial cells in response to hypoxia by HIF-1. TRAP1 (TNF receptor-associated protein 1), also designated HSP 75 is a mitochondrial matrix component that plays a role in the induction of apoptosis in response to reactive oxygen species.

Host Species: Rabbit

Species Reactivity: Human. Others not known.

Epitope: C-terminal

Immunogen: Synthetic peptide corresponding to C-terminal of human GRP94

Molecular Weight: 94 kDa

Positive Control: Breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen GRP94
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Clone Polyclonal
Concentration 200µg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C-terminal of human GRP94
Purification Method Breast CA
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.