Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ HDJ-2/DNAJ Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Catalog No. MS225R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS225R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS225R1 Supplier Epredia™ Supplier No. MS225R1

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments.

It is a novel human homolog of E. coli's DNAJ protein from heat shock protein 40 family. Hsp40 is involved in chaperonic protein folding HDJ-2 is cytoplasmic and nuclear membrane associated protein; upon heat shock it migrates and becomes associated with the Golgi complex, nucleolus and nuclear membrane HDJ-2 has a prenylated C-terminus and is not inducible upon heat shock.

Host Species: Mouse

Clone: KA2A5.6

Isotype: IgG1

Species Reactivity: Human, Monkey, Pig, Rabbit, Mouse, Rat, Hamster and Bacteria.

Epitope: aa 1-179

Immunogen: N-terminal purified recombinant fragment (aa 1-179) of HDJ-2 fusion protein

Molecular Weight: 44-47 kDa

Positive Control: MAD109, Raji cells, Tonsil

Cellular Localization: Cytoplasm (ground state) and Golgi complex, Nucleolus, and Nuclear membrane (upon heat shock)

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen HDJ-2/DNAJ Ab-1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone KA2A5.6
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen N-terminal purified recombinant fragment (aa 1-179) of HDJ-2 fusion protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Stress Response
Primary or Secondary Primary
Target Species Bacteria, Hamster, Human, Mouse, Non-human Primate, Pig, Rabbit, Rat
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.