Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Heat Shock Protein 27 (HSP27, Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotin, BSA and azide

Catalog No. MS101B1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS101B1 500 μL Biotin Concentrated 200μg/mL
MS101B0 100 μL Biotin Concentrated 200μg/mL
MS101P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS101PABX 200 μL Unconjugated Concentrated 1mg/mL
MS101P1 500 μL Unconjugated Concentrated 200μg/mL
MS101B 1 mL Biotin Concentrated 200μg/mL
MS101P 1 mL Unconjugated Concentrated 200μg/mL
MS101R1 1 mL Unconjugated Prediluted -
MS101R7 7 mL Unconjugated Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. MS101B1 Supplier Epredia™ Supplier No. MS101B1

Epredia™ Heat Shock Protein 27 (HSP27) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Hsp27 is identical to the estrogen-induced“p29” and“24K” protein. About 50% of breast carcinomas are positive for hsp27 especially those that are also positive for estrogen and/or progesterone receptor. HSP27 has also been implicated in drug resistance in cancer cells.

Host Species: Mouse

Clone: G3.1

Isotype: IgG1

Species Reactivity: Human, Chimpanzee and Monkey. Does not react with mouse and rat. Others not known.

Epitope: Not determined

Immunogen: Partially purified HSP27 (earlier called 24K) protein from breast cancer MCF-7 cells

Molecular Weight: 24-27 kDa

Positive Control: BT474 or MDA-MB-453 cells, Breast CA

Cellular Localization: Cytoplasmic/nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Heat Shock Protein 27 (HSP27) Ab-1
Applications Western Blot
Classification Monoclonal
Clone G3.1
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells
Quantity 500 μL
Regulatory Status RUO
Research Discipline Stress Response
Primary or Secondary Primary
Target Species Human, Non-human Primate
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.