Learn More
Epredia™ Lab Vision™ Heat Shock Protein 27 (HSP27, Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotin, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: G3.1
Isotype: IgG1
Species Reactivity: Human, Chimpanzee and Monkey. Does not react with mouse and rat. Others not known.
Epitope: Not determined
Immunogen: Partially purified HSP27 (earlier called 24K) protein from breast cancer MCF-7 cells
Molecular Weight: 24-27 kDa
Positive Control: BT474 or MDA-MB-453 cells, Breast CA
Cellular Localization: Cytoplasmic/nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Heat Shock Protein 27 (HSP27) Ab-1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | G3.1 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.