Learn More
Description
Host Species: Mouse
Clone: W27
Isotype: IgG2a
Species Reactivity: Human, Monkey and Cow. Others not known.
Epitope: Not determined
Immunogen: HSP70 protein from HeLa cells
Molecular Weight: 73 kDa (constitutive), 72 kDa (inducible)
Positive Control: BT474 or HeLa cells, Breast CA
Cellular Localization: Nuclear and cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Heat Shock Protein 70 (HSP70) Ab-2 |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | W27 |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | HSP70 protein from HeLa cells |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.