Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heat Shock Protein 70 (HSP70) Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS482P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS482P 1 mL Unconjugated Concentrated 200μg/mL
MS482B1 500 μL Biotin Concentrated 200μg/mL
MS482B 1 mL Biotin Concentrated 200μg/mL
MS482P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS482PABX 200 μL Unconjugated Concentrated 1mg/mL
MS482B0 100 μL Biotin Concentrated 200μg/mL
MS482P0 100 μL Unconjugated Concentrated 200μg/mL
MS482P1 500 μL Unconjugated Concentrated 200μg/mL
MS482R7 7 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. MS482P Supplier Epredia Supplier No. MS482P
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments.

Heat shock proteins (HSP's) or stress response proteins (SRP's) are synthesized in variety of environmental and pathophysiological stressful conditions. Many HSP's are involved in processes such as protein denaturation-renaturation, folding-unfolding, transport-translocation, activation-inactivation, and secretion. HSP70 is found to be associated with steroid receptors, actin, p53, polyoma T antigen, nucleotides, and other unknown proteins. Also, HSP70 has been shown to be involved in protective roles against thermal stress, cytotoxic drugs, and other damaging conditions.

Host Species: Mouse

Clone: W27

Isotype: IgG2a

Species Reactivity: Human, Monkey and Cow. Others not known.

Epitope: Not determined

Immunogen: HSP70 protein from HeLa cells

Molecular Weight: 73 kDa (constitutive), 72 kDa (inducible)

Positive Control: BT474 or HeLa cells, Breast CA

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Heat Shock Protein 70 (HSP70) Ab-2
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone W27
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen HSP70 protein from HeLa cells
Quantity 1 mL
Regulatory Status RUO
Research Discipline Stress Response
Primary or Secondary Primary
Target Species Bovine, Human, Non-human Primate
Form Concentrated
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.