Learn More
Heat Shock Protein 90α/hsp90α (HSP86) Ab-1 Rabbit Polyclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: aa 2-12
Immunogen: A synthetic 11-mer peptide, corresponding to aa 2-12 (PEETQTQDQPM-Cys) from the N-terminus end of mouse heat shock protein 90α (hsp90α)
Molecular Weight: 86 kDa
Positive Control: LS174T cells, MAD109 cells or breast CA
Cellular Localization: Cytoplasmic and nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Heat Shock Protein 90alpha/hsp90alpha (HSP86) Ab-1 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified w/o BSA and Azide |
| Host Species | Rabbit |
| Immunogen | A synthetic 11-mer peptide, corresponding to aa 2-12 (PEETQTQDQPM-Cys) from the N-terminus end of mouse heat shock protein 90alpha (hsp90alpha) |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.