Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heat Shock Protein 90α/hsp90α (HSP86) Ab-1 Rabbit Polyclonal Antibody, Epredia™

Catalog No. RB119R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Formulation:
Purified w/o BSA and Azide
Purified with BSA and Azide
Ready-to-use
Ready-to-use for IHC (UVLP)
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Formulation
RB119R7 7 mL Prediluted Ready-to-use
RB119P0 100 μL Concentrated Purified with BSA and Azide
RB119P1 500 μL Concentrated Purified with BSA and Azide
RB119P1ABX 500 μL Concentrated Purified w/o BSA and Azide
RB119P 1 mL Concentrated Purified with BSA and Azide
RB119PABX 1 mL Concentrated Purified w/o BSA and Azide
RB119R1 1 mL Prediluted Ready-to-use for IHC (UVLP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. RB119R7 Supplier Epredia Supplier No. RB119R7

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

Cells respond to heat and chemical stress by inducing the synthesis of a group of proteins referred to as heat shock proteins. HSP90 is present in cells at high levels in the cytoplasm and is composed of two separate gene products, HSP90α and HSP90Β.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 2-12

Immunogen: A synthetic 11-mer peptide, corresponding to aa 2-12 (PEETQTQDQPM-Cys) from the N-terminus end of mouse heat shock protein 90α (hsp90α)

Molecular Weight: 86 kDa

Positive Control: LS174T cells, MAD109 cells or breast CA

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Heat Shock Protein 90alpha/hsp90alpha (HSP86) Ab-1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Ready-to-use
Host Species Rabbit
Immunogen A synthetic 11-mer peptide, corresponding to aa 2-12 (PEETQTQDQPM-Cys) from the N-terminus end of mouse heat shock protein 90alpha (hsp90alpha)
Quantity 7 mL
Regulatory Status RUO
Research Discipline Stress Response
Primary or Secondary Primary
Target Species Human, Mouse
Form Prediluted
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.