Learn More
Epredia™ Lab Vision™ Hepatitis B Virus Core Antigen (HBVcAg) Ab-1 Rabbit Polyclonal Antibody with BSA and azide
Shop All Epredia ProductsDescription
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Hepatitis B virus
Positive Control: Hepatitis B virus infected liver
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Hepatitis B Virus Core Antigen (HBVcAg) Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Clone | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Hepatitis B virus |
| Quantity | 7 mL |
| Regulatory Status | ASR |
| Research Discipline | Infectious Disease |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.