Learn More
Epredia™ Lab Vision™ Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1 Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: T9; same as 3E7
Isotype: IgG1/λ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Purified HBVsAg, subtype ayw3
Positive Control: Hepatitis B virus infected liver
Cellular Localization: Cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | T9; same as 3E7 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Purified HbsAg, subtype ayw3 |
| Quantity | 1 mL |
| Regulatory Status | ASR |
| Research Discipline | Infectious Disease |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.