Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1 Mouse Monoclonal Antibody

Catalog No. MS314S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Supernatant
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS314S 1 mL Supernatant
MS314S1 500 μL Concentrated
MS314RQ 12 mL Ready-to-Use
MS314S0 100 μL Concentrated
MS314R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS314S Supplier Epredia™ Supplier No. MS314S
Add to Cart
Add to Cart

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1, Mouse Monoclonal Antibody.

Hepatitis B virus causes type B hepatitis and it belongs to hepadnavirus family. It is spherical in shape with a diameter of 42nm. It contains a 27nm partially double stranded DNA core enclosed within a lipoprotein coat. The antigenic activity of the nucleocapsid core is designated as hepatitis B core antigen. The antigens in the outer surface are called as hepatitis B virus surface antigens. Core antigens are localized within the nuclei whereas the surface antigens are present in the cytoplasm of the infected cells. Antibodies to surface antigens appear in circulation at an early stage of infection whereas the antibodies to the core antigens are detected after several weeks.

Host Species: Mouse

Clone: T9; same as 3E7

Isotype: IgG1/λ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified HBVsAg, subtype ayw3

Positive Control: Hepatitis B virus infected liver

Cellular Localization: Cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Hepatitis B Virus Surface Antigen (HBVsAg) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone T9; same as 3E7
Conjugate Unconjugated
Dilution 1:25-1:50 for 30 min at RT
Host Species Mouse
Immunogen Purified HbsAg, subtype ayw3
Quantity 1 mL
Regulatory Status ASR
Research Discipline Infectious Disease
Primary or Secondary Primary
Target Species Human
Content And Storage Store vial at 4deg.C. When stored at 2 to 8deg.C,
Form Supernatant
Isotype IgG1 λ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.