Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Hepatocyte Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1810R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1810R1 1 mL Prediluted
MS1810S0 100 μL Concentrated
MS1810S1 500 μL Concentrated
MS1810S 1 mL Concentrated
MS1810R7 7 mL Prediluted
MS1810RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS1810R1 Supplier Epredia™ Supplier No. MS1810R1

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Hepatocyte Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: OCH1E5

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Formalin-fixed failed human allograft liver that was mechanically disrupted

Positive Control: Liver CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Hepatocyte Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OCH1E5
Conjugate Unconjugated
Host Species Mouse
Immunogen Formalin fixed failed human allograft liver that was mechanically disrupted
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.