Learn More
Description
Host Species: Mouse
Clone: OCH1E5
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Formalin-fixed failed human allograft liver that was mechanically disrupted
Positive Control: Liver CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Hepatocyte Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | OCH1E5 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Formalin fixed failed human allograft liver that was mechanically disrupted |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.