Learn More
Description
Host Species: Rabbit
Clone: SP3
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Extracellular domain
Immunogen: Recombinant protein encoding extracellular domain of human HER-2
Molecular Weight: 185 kDa
Positive Control: Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | HER-2/c-erB-2/neu |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SP3 |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encoding extracellular domain of human HER-2 |
| Quantity | 7 mL |
| Regulatory Status | IVD |
| Research Discipline | Oncogenesis |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.