Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HER-2/c-erbB-2/neu Ab-16 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS600PA Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS600PA 1 mL 200μg/mL
MS600P0A 100 μL 200μg/mL
MS600P1A 500 μL 200μg/mL
MS600PABX 200 μL 1mg/mL
MS600P1ABX 100 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS600PA Supplier Epredia Supplier No. MS600PA
Add to Cart
Add to Cart

Provide accurate, reproducible results with the Epredia™ HER-2/c-erbB-2/neu Ab-16, Mouse Monoclonal Antibody.

c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Mouse

Clone: L26

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Involved in the formation of heterodimers

Immunogen: Extracellular domain of recombinant human HER-2/c-erbB-2 oncoprotein

Molecular Weight: 185kDa

Positive Control: SKBR-3 cells

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Not suitable)
  • Immunoprecipitation (Native only)
  • Inhibits Binding of EGF (∽70%), NDF (∽40%)
  • Reduces Affinity of EGF (3-fold) and NDF (2-fold)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen HER-2/c-erbB-2/neu Ab-16
Applications Immunoprecipitation
Classification Monoclonal
Clone L26
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Extracellular domain of recombinant human c-erbB-2/HER-2 oncoprotein
Quantity 1 mL
Regulatory Status RUO
Research Discipline EGFR Family
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.