Learn More
Description
Host Species: Mouse
Clone: 9G6.10
Isotype: IgG1
Species Reactivity: Human. Does not react with mouse and rat. Others not tested.
Epitope: Extracellular domain
Immunogen: RAC311 cells transfected with SV40-neu construct, expressing human neu protein
Molecular Weight: 185kDa
Positive Control: SKBR-3 cells, or Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Frozen only) Immunofluorescence
- Immunoprecipitation
- Flow Cytometry
- Inhibits Colony Formation
- Partially Inhibits Tyrosine Kinase Activity
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | HER-2/c-erbB-2/neu Ab-2 |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 9G6.10 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | RAC311 cells transfected with SV40-neu construct, expressing human neu protein |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.