Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HER-2/c-erbB-2/neu Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS229PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS229PABX 200 μL 1mg/mL
MS229P0 100 μL 200μg/mL
MS229P1 500 μL 200μg/mL
MS229P 1 mL 200μg/mL
MS229P1ABX 100 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS229PABX Supplier Epredia Supplier No. MS229PABX

Provide accurate, reproducible results with the Epredia™ HER-2/c-erbB-2/neu Ab-2, Mouse Monoclonal Antibody.

c-erbB-2, second member (c-erbB-2/HER-2/neu) of the c-erbB family is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Mouse

Clone: 9G6.10

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not tested.

Epitope: Extracellular domain

Immunogen: RAC311 cells transfected with SV40-neu construct, expressing human neu protein

Molecular Weight: 185kDa

Positive Control: SKBR-3 cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen only) Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • Inhibits Colony Formation
  • Partially Inhibits Tyrosine Kinase Activity

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen HER-2/c-erbB-2/neu Ab-2
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 9G6.10
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen RAC311 cells transfected with SV40-neu construct, expressing human neu protein
Quantity 200 μL
Regulatory Status RUO
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.