Learn More
Description
Host Species: Mouse
Clone: N24
Isotype: IgG1
Species Reactivity: Human. Does not react with mouse and rat. Others not tested.
Epitope: Extracellular domain
Immunogen: Intact SKBR-3 breast cancer cells
Molecular Weight: 185 kDa
Positive Control: SKBR-3 cells, or Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunofluorescence
- Immunoprecipitation
- Accelerates (∽50%) HER-2 Internalization
- Inhibits (∽80%) Tumorigenic Cell Growth
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | HER-2/c-erbB-2/neu Ab-5 |
| Applications | Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | N24 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Intact SKBR-3 breast cancer cells |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.