Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HER-2/c-erbB-2/neu Ab-8 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS325B Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS325B 1 mL Biotin 200μg/mL
MS325B0 100 μL Biotin 200μg/mL
MS325B1 500 μL Biotin 200μg/mL
MS325P1 500 μL Unconjugated 200μg/mL
MS325P1ABX 100 μL Unconjugated 1mg/mL
MS325P0 100 μL Unconjugated 200μg/mL
MS325P 1 mL Unconjugated 200μg/mL
MS325PABX 200 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS325B Supplier Epredia Supplier No. MS325B
Add to Cart
Add to Cart

Recommended for Immunohistochemistry, Western Blotting and Immunoprecipitation (Native and Denatured), Epredia™ HER-2/c-erbB-2/neu Ab-8, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-erbB-2 is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein. The c-erbB-2 protein is overexpressed in a variety of carcinomas especially those of breast and ovary.

Host Species: Mouse

Clone: e2-4001

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: Cytoplasmic domain

Immunogen: Cytoplasmic domain of recombinant human HER-2/c-erbB-2 oncoprotein

Molecular Weight: 185 kDa

Positive Control: SKBR-3 cells, or T47D cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunoprecipitation (Native and Denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen HER-2/c-erbB-2/neu Ab-8
Applications Immunoprecipitation
Classification Monoclonal
Clone e2-4001
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.