Learn More
Description
Host Species: Mouse
Clone: e2-4001
Isotype: IgG1
Species Reactivity: Human, Mouse and Rat. Others not tested.
Epitope: Cytoplasmic domain
Immunogen: Cytoplasmic domain of recombinant human HER-2/c-erbB-2 oncoprotein
Molecular Weight: 185 kDa
Positive Control: SKBR-3 cells, or T47D cells, or Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry
- Western Blotting
- Immunoprecipitation (Native and Denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | HER-2/c-erbB-2/neu Ab-8 |
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Clone | e2-4001 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.