Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ HER-2/c-erbB-2/neu (Phospho-specific) Ab-18 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Catalog No. MS1072B Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS1072B 1 mL Biotin 200μg/mL
MS1072B1 500 μL Biotin 200μg/mL
MS1072P0 100 μL Unconjugated 200μg/mL
MS1072P1 500 μL Unconjugated 200μg/mL
MS1072P 1 mL Unconjugated 200μg/mL
MS1072P1ABX 100 μL Unconjugated 1mg/mL
MS1072PABX 200 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1072B Supplier Epredia™ Supplier No. MS1072B

Provide accurate, reproducible results with the Epredia™ HER-2/c-erbB-2/neu (Phospho-specific) Ab-18, Mouse Monoclonal Antibody.

c-erbB-2 (HER-2/neu) is a 185 kDa receptor tyrosine kinase belonging to the epidermal growth factor receptor family, which also includes EGFR (HER-1), c-erbB-3 (HER-3), and c-erbB-4 (HER-4). Amplification of the HER-2 gene with overexpression of the protein occurs in 20-30% of breast cancers, and tumor progression in mammary cancer is associated with elevated levels of tyrosine-phosphorylated neu and erbB-3. Tyr1248 is the phosphorylation site that is most tightly linked to oncogenic transformation and coupling to the ras/MAP kinase signaling pathway. Cases of invasive breast carcinoma with the phosphorylated receptor have displayed aggressive clinicopathological features and adverse prognoses. Activation of HER-2 is frequent in ductal carcinoma in situ suggesting that HER-2 signaling plays a critical role in the early stages of breast tumorigenesis.

Host Species: Mouse

Clone: PN2A

Isotype: IgG1

Species Reactivity: Human and Rat. Others not tested.

Epitope: aa 1242-1255 with pTyr

Immunogen: Tyrosine-phosphorylated synthetic peptide corresponding to aa 1242-1255 [Lys-TAENPE-pY-LGLDVPV] from the C-terminus of human HER-2 protein (P-Tyr1248)

Molecular Weight: 185 kDa

Positive Control: EGF-treated SKBR-3 cells. About 5% of breast CA are positive for phospho-HER-2/c-erbB-2/neu oncoprotein.

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Not verified)
  • Flow Cytometry (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen HER-2/c-erbB-2/neu (Phospho-specific) Ab-18
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PN2A
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen Tyrosine-phosphorylated synthetic peptide corresponding to aa1242-1255 [Lys-TAENPE-pY-LGLDVPV] from the C-terminus of human c-erbB-2 protein (P-Tyr1248)
Quantity 1 mL
Regulatory Status RUO
Research Discipline EGFR Family
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.