Learn More
Epredia™ Lab Vision™ HER-2/c-erbB-2/neu (Phospho-specific) Ab-18 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: PN2A
Isotype: IgG1
Species Reactivity: Human and Rat. Others not tested.
Epitope: aa 1242-1255 with pTyr
Immunogen: Tyrosine-phosphorylated synthetic peptide corresponding to aa 1242-1255 [Lys-TAENPE-pY-LGLDVPV] from the C-terminus of human HER-2 protein (P-Tyr1248)
Molecular Weight: 185 kDa
Positive Control: EGF-treated SKBR-3 cells. About 5% of breast CA are positive for phospho-HER-2/c-erbB-2/neu oncoprotein.
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Not verified)
- Flow Cytometry (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | HER-2/c-erbB-2/neu (Phospho-specific) Ab-18 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | PN2A |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Tyrosine-phosphorylated synthetic peptide corresponding to aa1242-1255 [Lys-TAENPE-pY-LGLDVPV] from the C-terminus of human c-erbB-2 protein (P-Tyr1248) |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.