Learn More
Description
Host Species: Mouse
Clone: 3B5
Isotype: IgG1
Species Reactivity: Human, Monkey, Mouse and Rat. Others not tested.
Epitope: C-terminus
Immunogen: A synthetic peptide from the C-terminus of human HER-2 protein. This sequence is identical in rat neu protein.
Molecular Weight: 185 kDa
Positive Control: SKBR-3 or MAD109 cells, or Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry
- Western Blotting
- Immunoprecipitation (Native and Denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | HER-2/c-erbB-2/neu5 Ab-15 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 3B5 |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | A synthetic peptide from the C-terminus of human c-erbB-2 protein. This Sequence Is Identical In Rat Neu Protein. |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.