Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HER-2/c-erbB-2/neu5 Ab-15 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS599B0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS599B0 100 μL Biotin 200μg/mL
MS599B1 500 μL Biotin 200μg/mL
MS599B 1 mL Biotin 200μg/mL
MS599P1ABX 100 μL Unconjugated 1mg/mL
MS599P1 500 μL Unconjugated 200μg/mL
MS599P0 100 μL Unconjugated 200μg/mL
MS599PABX 200 μL Unconjugated 1mg/mL
MS599P 1 mL Unconjugated 200μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS599B0 Supplier Epredia Supplier No. MS599B0
Add to Cart
Add to Cart

Recommended for Immunohistochemistry, Western Blotting and Immunoprecipitation (Native and Denatured), Epredia™ HER-2/c-erbB-2/neu5 Ab-15, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Mouse

Clone: 3B5

Isotype: IgG1

Species Reactivity: Human, Monkey, Mouse and Rat. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide from the C-terminus of human HER-2 protein. This sequence is identical in rat neu protein.

Molecular Weight: 185 kDa

Positive Control: SKBR-3 or MAD109 cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunoprecipitation (Native and Denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen HER-2/c-erbB-2/neu5 Ab-15
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3B5
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen A synthetic peptide from the C-terminus of human c-erbB-2 protein. This Sequence Is Identical In Rat Neu Protein.
Quantity 100 μL
Regulatory Status RUO
Research Discipline EGFR Family
Primary or Secondary Primary
Target Species Human, Mouse, Non-human Primate, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.