Learn More
Description
Host Species: Mouse
Clone: 2F12
Isotype: IgG2a/κ
Species Reactivity: Human, Cow, Rat and Mouse. Others not tested.
Epitope: aa 1295-1323
Immunogen: Recombinant rat HER-3/c-erbB-3 oncoprotein
Molecular Weight: 185kDa
Positive Control: T47D cells
Cellular Localization: Cytoplasmic and membrane
Recommended for:
- Western Blotting
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | HER-3/c-erbB-3 Ab-2 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 2F12 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant rat c-erbB-3/HER-3 oncoprotein |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.