Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HER-3/c-erbB-3 Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS201P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS201P0 100 μL 200μg/mL
MS201P1 500 μL 200μg/mL
MS201P 1 mL 200μg/mL
MS201P1ABX 100 μL 1mg/mL
MS201PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS201P0 Supplier Epredia Supplier No. MS201P0
Add to Cart
Add to Cart

Epredia™ HER-3/c-erbB-3 Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-erbB-3/HER-3 is the third member of the type I family of growth factor receptors. It binds to ligands in the heregulin family. c-erbB-3 is over-expressed in a variety of tumors including breast, stomach, pancreas, and colon. Heregulin & EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents.

Host Species: Mouse

Clone: 2F12

Isotype: IgG2a/κ

Species Reactivity: Human, Cow, Rat and Mouse. Others not tested.

Epitope: aa 1295-1323

Immunogen: Recombinant rat HER-3/c-erbB-3 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D cells

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen HER-3/c-erbB-3 Ab-2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2F12
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant rat c-erbB-3/HER-3 oncoprotein
Quantity 100 μL
Regulatory Status RUO
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.