Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HER-3/c-erbB-3 Ab-4 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS262P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS262P0 100 μL Unconjugated 200μg/mL
MS262P1 500 μL Unconjugated 200μg/mL
MS262P 1 mL Unconjugated 200μg/mL
MS262B0 100 μL Biotin 200μg/mL
MS262B1 500 μL Biotin 200μg/mL
MS262B 1 mL Biotin 200μg/mL
MS262P1ABX 100 μL Unconjugated 1mg/mL
MS262PABX 200 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS262P0 Supplier Epredia Supplier No. MS262P0

Epredia™ HER-3/c-erbB-3 Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

c-erbB-3/HER-3 is the third member of the type I family of growth factor receptors. It binds to ligands in the heregulin family. c-erbB-3 is over-expressed in a variety of tumors including breast, stomach, pancreas, and colon. Heregulin & EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents.

Host Species: Mouse

Clone: H3.90.6; same as Ab90

Isotype: IgG2a

Species Reactivity: Human. Others not tested.

Epitope: Extracellular domain

Immunogen: Extracellular fragment of recombinant human HER-3/c-erbB-3 oncoprotein

Molecular Weight: 185kDa

Positive Control: T47D cells

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • Effect on Heregulin (NDF) Binding (Noninhibitory)
  • Effect on Tumor Growth (Noninhibitory)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen HER-3/c-erbB-3 Ab-4
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone H3.90.6; same as Ab90
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Extracellular fragment of recombinant human c-erbB-3/HER-3 oncoprotein
Quantity 100 μL
Regulatory Status RUO
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.