Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ HER-4/c-erbB-4 Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB9045R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB9045R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RB9045R1 Supplier Epredia™ Supplier No. RB9045R1

Recommended for Immunohistochemistry (Formalin/paraffin) and Western Blotting, Epredia™ HER-4/c-erbB-4, Rabbit Polyclonal Antibody provides accurate, reproducible results.

c-erbB-4 is the fourth member of class I receptor kinase family. HER-4 is most predominantly expressed in several breast carcinoma cell lines, and in normal skeletal muscle, heart, pituitary, brain, and cerebellum. Breast tumor cell lines T47-D, MDA-MB-453, BT-474 and H3396 are found to have the highest levels of mRNA, and intermediate levels are seen in MCF-7, MDA-MB-330 and MDA-MB-361. Expression of erbB-4 is low or absent in some breast tumor cell lines such as MDA-MB-231, MDA-MB-157, MDA-MB-468, and SKBR-3.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of human HER-4 protein

Molecular Weight: 185kDa

Positive Control: Breast CA T47D cells

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen HER-4/c-erbB-4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Rabbit
Immunogen A synthetic peptide from C-terminus of human c-erbB-4 protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline EGFR Family
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.