Learn More
Epredia™ Lab Vision™ Heregulin/NDF/GGF/Neuregulin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: 7D5
Isotype: IgG2a
Species Reactivity: Human, Rat and Mouse.
Epitope: Extracellular
Immunogen: Recombinant extracellular domain of rat NDF protein
Molecular Weight: 44kDa
Positive Control: HRGα1 or HRGβ1 recombinant protein, Prostate CA
Cellular Localization: Cytoplasmic and intercellular spaces
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Heregulin/NDF/GGF/Neuregulin Ab-1 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 7D5 |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | Recombinant extracellular domain of rat NDF protein |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.