Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Heregulin/NDF/GGF/Neuregulin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Catalog No. MS272B1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS272B1 500 μL Concentrated
MS272B0 100 μL Concentrated
MS272B 1 mL Concentrated
MS272R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS272B1 Supplier Epredia™ Supplier No. MS272B1

Recommended for Western Blotting and Immunoprecipitation, Epredia™ Heregulin/NDF/GGF/Neuregulin Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Heregulin (HRG) or neu differentiation factor (NDF) binds to c-erbB-3 and c-erbB-4 receptors on mammary and neuronal cells with low and high affinities, respectively. HRG/NDF exists in several isoforms, which are classified in two groups, α and β that differ in their EGF-like domain and in receptor binding affinity.

Host Species: Mouse

Clone: 7D5

Isotype: IgG2a

Species Reactivity: Human, Rat and Mouse.

Epitope: Extracellular

Immunogen: Recombinant extracellular domain of rat NDF protein

Molecular Weight: 44kDa

Positive Control: HRGα1 or HRGβ1 recombinant protein, Prostate CA

Cellular Localization: Cytoplasmic and intercellular spaces

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Heregulin/NDF/GGF/Neuregulin Ab-1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7D5
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen Recombinant extracellular domain of rat NDF protein
Quantity 500 μL
Regulatory Status RUO
Research Discipline EGFR Family
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Concentrated
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.