Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ HIER Buffer M (100X)

Catalog No. ta135hbm Shop All Epredia Products
Click to view available options
Quantity:
9 x 15 mL

Perform heat induced epitope retrieval (HIER) on formalin-fixed paraffin-embedded tissue sections using the Epredia™ Lab Vision™ HIER Buffer M (100X)

  • For in vitro diagnostic use
  • Store at room temperature

Specifications

Content And Storage Room temperature
Dilution 100X
Product Type HIER
Quantity 9 x 15 mL
For Use With (Application) Immunohistochemistry
pH 6 to 7
Regulatory Status IVD
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.