Learn More
Epredia™ Lab Vision™ Human Papilloma Virus (HPV) Ab-3 Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: K1H8
Isotype: IgG1/κ
Species Reactivity: Human Papilloma Virus type 6, 11, 16, 18, 31, 33, 42, 51, 52, 56 and 58.
Epitope: Nonconfirmational internal linear epitope of major capsid protein
Immunogen: Alkaline disrupted HPV type 1
Molecular Weight: 57kDa (major band)
Positive Control: Infected cells and tissue
Cellular Localization: Nuclei of infected cells
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Human Papilloma Virus (HPV) Ab-3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | K1H8 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Alkaline disrupted HPV type 1 |
| Quantity | 12 mL |
| Regulatory Status | ASR, IVD |
| Research Discipline | Infectious Disease |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.