Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Human Papilloma Virus (HPV) Ab-3 Mouse Monoclonal Antibody

Catalog No. MS1826S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS1826S1 500 μL
MS1826S 1 mL
MS1826R7 7 mL
MS1826S0 100 μL
MS1826RQ 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1826S1 Supplier Epredia™ Supplier No. MS1826S1A
Add to Cart
Add to Cart

Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Human Papilloma Virus (HPV) Ab-3, Mouse Monoclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Human papilloma virus is non-enveloped double stranded circular DNA virus with icosahedral capsid.

Host Species: Mouse

Clone: K1H8

Isotype: IgG1/κ

Species Reactivity: Human Papilloma Virus type 6, 11, 16, 18, 31, 33, 42, 51, 52, 56 and 58.

Epitope: Nonconfirmational internal linear epitope of major capsid protein

Immunogen: Alkaline disrupted HPV type 1

Molecular Weight: 57kDa (major band)

Positive Control: Infected cells and tissue

Cellular Localization: Nuclei of infected cells

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Human Papilloma Virus (HPV) Ab-3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone K1H8
Conjugate Unconjugated
Host Species Mouse
Immunogen Alkaline disrupted HPV type 1
Purification Method Infected cells and tissue
Quantity 500 μL
Regulatory Status ASR, IVD
Research Discipline Infectious Disease
Primary or Secondary Primary
Target Species Human, Virus
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.