Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Sodium Iodide Symporter (hNIS) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1653P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1653P 1 mL Concentrated 200μg/mL
MS1653P0 100 μL Concentrated 200μg/mL
MS1653P1ABX 100 μL Concentrated 1mg/mL
MS1653PABX 200 μL Concentrated 1mg/mL
MS1653P1 500 μL Concentrated 200μg/mL
MS1653R1 1 mL Prediluted Prediluted
MS1653R7 7 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1653P Supplier Epredia Supplier No. MS1653P

Provide accurate, reproducible results with the Epredia™ Human Sodium Iodide Symporter (hNIS) Ab-1, Mouse Monoclonal Antibody.

Human Sodium Iodide Symporter (hNIS) is responsible for iodide concentrating ability within thyroid follicular cells. It is a membrane bound glycoprotein with 13 membrane spanning domains and 14 extramembranous domains. It may represent an autoantigen in thyroid. Antibody to NIS showed strong immunostaining in Graves tissues, intermediate staining in papillary and follicular thyroid cancer, and no staining in Hurtle cell cancer or in nonthyroidal tissue.

Host Species: Mouse

Clone: FP5A

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse and rat. Others not tested.

Epitope: aa 625-643

Immunogen: A synthetic peptide corresponding to aa 625-643 of hNIS protein

Molecular Weight: 97kDa

Positive Control: Thyroid

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Human Sodium Iodide Symporter (hNIS)
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone FP5A
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen A synthetic peptide corresponding to aa625-643 of hNIS protein
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.