Learn More
Human Sodium Iodide Symporter (hNIS) Ab-1 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: FP5A
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with mouse and rat. Others not tested.
Epitope: aa 625-643
Immunogen: A synthetic peptide corresponding to aa 625-643 of hNIS protein
Molecular Weight: 97kDa
Positive Control: Thyroid
Cellular Localization: Membrane and cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Human Sodium Iodide Symporter (hNIS) Ab-1 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | FP5A |
| Concentration | Prediluted |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | A synthetic peptide corresponding to aa625-643 of hNIS protein |
| Quantity | 7 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.