Learn More
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Helicobacter pylori
Epitope: Not determined
Immunogen: Purified Helicobacter pylori
Positive Control: Helicobacter pylori infected tissue
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Helicobacter pylori |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Clone | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Purified Helicobacter pylori |
| Quantity | 12 mL |
| Regulatory Status | IVD |
| Research Discipline | Infectious Disease |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.