Learn More
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: IgD isolated from a pool of normal human sera
Molecular Weight: 50kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | IgD (delta-Heavy Chain) Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Clone | Polyclonal |
| Host Species | Rabbit |
| Immunogen | IgD isolated from a pool of normal human sera |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Research Discipline | Immunoglobulins |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.