Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF-1R (α-subunit) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS641B0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS641B0 100 μL Biotin 200μg/mL
MS641B1 500 μL Biotin 200μg/mL
MS641P1 500 μL Unconjugated 200μg/mL
MS641B 1 mL Biotin 200μg/mL
MS641P0 100 μL Unconjugated 200μg/mL
MS641P 1 mL Unconjugated 200μg/mL
MS641P1ABX 100 μL Unconjugated 1mg/mL
MS641PABX 200 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS641B0 Supplier Epredia Supplier No. MS641B0

Provide accurate, reproducible results with the Epredia™ IGF-1R (α-Subunit) Ab-1, Mouse Monoclonal Antibody.

IGF-1R (Insulin-like Growth Factor-1 Receptor) consists of α- and Β-subunits, which are disulfide-linked in a Β-α-α-Β configuration in the mature receptor. The α-subunit is completely extracellular, while the Β-subunit spans the membrane and the intracellular portion has intrinsic tyrosine kinase activity.

Host Species: Mouse

Clone: 24-31

Isotype: IgG1

Species Reactivity: Human. Weakly reacts with Rabbit. Does not react with rat. Pig unknown.

Epitope: aa 283-440 (exon 4-6)

Immunogen: IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA

Molecular Weight: 125kDa

Positive Control: Placenta or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Western Blotting (Not suitable)
  • Immunoprecipitation
  • ELISA
  • Acts as a Weak IGF-like Agonist in Bioassays (Thymidine incorporation into DNA)
  • Does Not Significantly Inhibit IGF-1 Binding

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen IGF-1R (alpha-Subunit) Ab-1
Applications Immunoprecipitation
Classification Monoclonal
Clone 24-31
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.