Learn More
Description
Host Species: Mouse
Clone: 24-31
Isotype: IgG1
Species Reactivity: Human. Weakly reacts with Rabbit. Does not react with rat. Pig unknown.
Epitope: aa 283-440 (exon 4-6)
Immunogen: IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA
Molecular Weight: 125kDa
Positive Control: Placenta or Breast CA
Cellular Localization: Membrane
Recommended for:
- Western Blotting (Not suitable)
- Immunoprecipitation
- ELISA
- Acts as a Weak IGF-like Agonist in Bioassays (Thymidine incorporation into DNA)
- Does Not Significantly Inhibit IGF-1 Binding
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | IGF-1R (alpha-Subunit) Ab-1 |
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 24-31 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.