Learn More
Description
Host Species: Mouse
Clone: 24-60
Isotype: IgG2a/κ
Species Reactivity: Human. Weakly reacts with Rabbit and Pig (Ab-2 is better). Does not react with rat.
Epitope: aa 184-283
Immunogen: IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA
Molecular Weight: 125kDa
Positive Control: Placenta or Breast CA
Cellular Localization: Membrane Ab-4 is excellent for immunoprecipitation of alpha-subunit of IGF-1R.
Recommended for:
- Western Blotting (Not suitable)
- Immunoprecipitation
- Acts as a Weak IGF-like Agonist in Bioassays (Thymidine incorporation into DNA)
- Substantially (>80%) Inhibits IGF-1 Binding
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | IGF-1R (alpha-Subunit) Ab-4 |
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 24-60 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.