Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ IgM (μ-Heavy Chain) Ab-2 Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB1434A0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB1434A0 100 μL Concentrated
RB1434A1 500 μL Concentrated
RB1434A 1 mL Concentrated
RB1434R1 1 mL Prediluted
RB1434R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RB1434A0 Supplier Epredia™ Supplier No. RB1434A0

IgM (μ-Heavy Chain) Ab-2, Rabbit PAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: IgM isolated from human serum of a patient with Waldenstrom's

Molecular Weight: 50kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic Ab-2 recognizes the μ heavy chain of human immunoglobulins. Ab-2 does not cross-react with γ (IgG), α (IgA), ε (IgE), or δ (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. Ab-2 is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen IgM (µ-Heavy Chain) Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen IgM isolated from human serum of a patient with Waldenstroms
Quantity 100 μL
Regulatory Status IVD
Research Discipline Immunoglobulins
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.