Learn More
Epredia™ Lab Vision™ IgM (μ-Heavy Chain) Ab-2 Rabbit Polyclonal Antibody with BSA and azide
Shop All Epredia ProductsDescription
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not known.
Immunogen: IgM isolated from human serum of a patient with Waldenstrom's
Molecular Weight: 50kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic Ab-2 recognizes the μ heavy chain of human immunoglobulins. Ab-2 does not cross-react with γ (IgG), α (IgA), ε (IgE), or δ (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. Ab-2 is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | IgM (µ-Heavy Chain) Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Clone | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | IgM isolated from human serum of a patient with Waldenstroms |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Research Discipline | Immunoglobulins |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.