Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Insulin Ab-6 (INS04 + INS05) Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1379R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1379R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS1379R1 Supplier Epredia™ Supplier No. MS1379R1

Epredia™ Insulin Ab-6 (INS04 + INS05), Mouse Monoclonal Antibody provides accurate, reproducible results.

Insulin is a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as Β-cell of islets of Langerhans and tumor (insulinoma) marker.

Host Species: Mouse

Clone: INS04 + INS05

Isotype: IgG1/κ + IgG1/κ

Species Reactivity: Human, Rat and Pig. Others not tested.

Epitope: Not determined

Immunogen: Insulin protein

Molecular Weight: 6kDa

Positive Control: Pancreas

Cellular Localization: Cytoplasmic Ab-6 cocktail is especially designed for sensitive detection of insulin .

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Insulin Ab-6 (INS04 plus INS05)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone INS04 + INS05
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Insulin protein
Quantity 1 mL
Regulatory Status IVD
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human, Pig, Rat
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.