Learn More
Description
Host Species: Mouse
Clone: 83-14
Isotype: IgG2a/κ
Species Reactivity: Human. Weakly with Cow, Pig and Sheep. Does not react with rabbit, mouse and rat. Others not known.
Epitope: aa 469-592 (exon 7/8)
Immunogen: IM-9 lymphocytes followed by purified insulin receptor
Molecular Weight: 135kDa
Positive Control: IM-9 lymphocyte cells, Placenta
Cellular Localization: Membrane Ab-1 inhibits insulin binding and mimics insulin action. It is excellent for immunoprecipitation of alpha-subunit of IR.
Recommended for:
- Immunoprecipitation
- ELISA
- Insulin-like Agonist
- Inhibits (∽80%) Insulin Binding (10nM)
- Tyrosine Kinase Assay (Ab-mediated capture on microplates)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Insulin Receptor (alpha-Subunit) Ab-1 |
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 83-14 |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | IM-9 lymphocytes followed by purified insulin receptor |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.