Learn More
Description
Host Species: Mouse
Clone: 83-7
Isotype: IgG1/κ
Species Reactivity: Human, Cow, Pig, Sheep and Rabbit. Does not react with mouse and rat.
Epitope: Exon 3
Immunogen: IM-9 lymphocytes followed by IR
Molecular Weight: 135kDa
Positive Control: IM-9 lymphocyte cells, Placenta
Cellular Localization: Cell membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- ELISA
- Flow Cytometry
- Insulin-like Agonist
- Does Not Significantly Inhibit Insulin Binding
- Tyrosine Kinase Assay (Ab-mediated capture on microplates)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Insulin Receptor (alpha-Subunit) Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 83-7 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | IM-9 lymphocytes followed by IR |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.