Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin Receptor (α-subunit) Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS632PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS632PABX 200 μL Unconjugated Concentrated 1mg/mL
MS632P1 500 μL Unconjugated Concentrated 200μg/mL
MS632B0 100 μL Biotin Concentrated 200μg/mL
MS632B 1 mL Biotin Concentrated 200μg/mL
MS632P0 100 μL Unconjugated Concentrated 200μg/mL
MS632P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS632P 1 mL Unconjugated Concentrated 200μg/mL
MS632R1 1 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS632PABX Supplier Epredia Supplier No. MS632PABX

Epredia™ Insulin Receptor (α-Subunit) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain, whereas the α-subunit (135kDa) is completely extracellular.

Host Species: Mouse

Clone: 83-7

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Pig, Sheep and Rabbit. Does not react with mouse and rat.

Epitope: Exon 3

Immunogen: IM-9 lymphocytes followed by IR

Molecular Weight: 135kDa

Positive Control: IM-9 lymphocyte cells, Placenta

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • ELISA
  • Flow Cytometry
  • Insulin-like Agonist
  • Does Not Significantly Inhibit Insulin Binding
  • Tyrosine Kinase Assay (Ab-mediated capture on microplates)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Insulin Receptor (alpha-Subunit) Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 83-7
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen IM-9 lymphocytes followed by IR
Quantity 200 μL
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Bovine, Human, Pig, Rabbit, Sheep
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.