Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin Receptor (β-subunit) Ab-5 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS635P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS635P0 100 μL 200μg/mL
MS635P1 500 μL 200μg/mL
MS635P 1 mL 200μg/mL
MS635P1ABX 100 μL 1mg/mL
MS635PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS635P0 Supplier Epredia Supplier No. MS635P0

Epredia™ Insulin Receptor (β-Subunit) Ab-5, Mouse Monoclonal Antibody provides accurate, reproducible results.

The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain with tyrosine kinase acivity, whereas the α-subunit (135kDa) is completely extracellular.

Host Species: Mouse

Clone: CT-1

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat.

Epitope: aa-GRVLTLPRS- within the C-terminus

Immunogen: A 15-mer synthetic peptide corresponding to aa Tyr-KKNGRILTLPRSNPS from the C-terminal of human insulin receptor β-subunit

Molecular Weight: 95kDa

Positive Control: IM-9 lymphocyte cells

Cellular Localization: Membrane Ab-5 reacts with autophosphorylated and unoccupied receptors as well as the receptor-insulin complex. Ab-5 does not affect insulin-stimulated autophosphorylation.

Recommended for:

  • Immunoprecipitation
  • Affinity Purification (With peptide elution)
  • ELISA (As capture antibody only)
  • Tyrosine Kinase Assay (Ab-mediated capture on microplates)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Insulin Receptor (beta-Subunit) Ab-5
Applications Immunoprecipitation
Classification Monoclonal
Clone CT-1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen A 15-mer synthetic peptide corresponding to aa Tyr-KKNGRILTLPRSNPS from the C-terminal of human insulin receptor Beta-subunit
Quantity 100 μL
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.